96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 1
Tired of paying too much for ECM plates? Try these ECM plates that are easy on your budget »
96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 1
Thermo Scientific™
96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 1
Green features
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate.
Have Questions?
Catalog number 267576
Price (USD)/ Case of 50
92.25
-
Add to cart
Price (USD)/ Case of 50
92.25
Add to cart
96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 1
Catalog number267576
Price (USD)/ Case of 50
92.25
-
Add to cart
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
Culture Area0.35 cm2/well
Array8 x 12
Packaging1/Pk., 50/Cs.
Product LineNunc
ColorClear
Product TypeMicroplate
Skirt StyleLow Flange
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Green FeaturesLess waste, fewer resources used
No. per Case50
No. per Pack1
Unit SizeCase of 50