Palladium, 99.9%, (trace metal basis), sponge, Thermo Scientific Chemicals
Palladium, 99.9%, (trace metal basis), sponge, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 99.9%, (trace metal basis), sponge, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 268150010
Price (USD)/ Each
318.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
318.00
Add to cart
Palladium, 99.9%, (trace metal basis), sponge, Thermo Scientific Chemicals
Catalog number268150010
Price (USD)/ Each
318.00
-
Add to cart
Chemical Identifiers
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
Molecular Weight (g/mol)106.42
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey
Total trace metal impurities=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only