Acetonitrile, 99.8%, for HPLC, Thermo Scientific Chemicals
Acetonitrile, 99.8%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, 99.8%, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Catalog number 268270010
Price (USD)/ Each
170.65
Online exclusive
200.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
200.00 
Save 29.35 (15%)
Add to cart
Acetonitrile, 99.8%, for HPLC, Thermo Scientific Chemicals
Catalog number268270010
Price (USD)/ Each
170.65
Online exclusive
200.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
Molecular Weight (g/mol)41.05
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.0002 %
UVat 240 nm A: =<0.015
UVat 260 nm A: =<0.005
UVat 280 nm A: =<0.005
Filtered to 0.2 micrometerFiltered
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only