1x1 image pixel for data collection
Water, for HPLC gradient grade
Water, for HPLC gradient grade
Water, for HPLC gradient grade
Water, for HPLC gradient grade
Thermo Scientific Chemicals

Water, for HPLC gradient grade

CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
4 L
Catalog number 268300025
Price (USD)
114.00
Each
In stock
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
114.00
Each
Add to cart
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Gradient analysis:
Appearance (Form)Liquid
Residue after evaporation=<0.0005 %
Absorbance largest eluted peak=<0.002 A.U.
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection