Thermo Scientific Chemicals

Water, for HPLC gradient grade, Thermo Scientific Chemicals

Catalog number: 268300040
4 L, Each
Thermo Scientific Chemicals

Water, for HPLC gradient grade, Thermo Scientific Chemicals

Catalog number: 268300040
4 L, Each
Quantity
Catalog number: 268300040
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
Appearance (Form)
Liquid
Gradient analysis
:
Appearance (Color)
Clear colorless
Residue after evaporation
=<0.0005 %
Absorbance largest eluted peak
=<0.002 A.U.

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text