Thermo Scientific Chemicals

Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals

Catalog number: 268350040
4 L, Each
Thermo Scientific Chemicals

Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals

Catalog number: 268350040
4 L, Each
Quantity
Catalog number: 268350040
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Color scale
=<10 APHA
Acidity
=<0.0015 meq/g
UV
1 cm cell vs HPLC-grade water
Appearance (Form)
Clear liquid
Water
=<0.03 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text