Thermo Scientific Chemicals

Potassium bromate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 268392500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium bromate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 268392500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 268392500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-01-2
IUPAC Name
potassium bromate
Molecular Formula
BrKO3
InChI Key
OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES
[K+].[O-][Br](=O)=O
Heavy metals (as Pb)
=<5 ppm
Sulfate (SO4)
=<50 ppm
Sodium (Na)
=<100 ppm
Appearance (Color)
White
Loss on drying
=<0.1 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text