Benzyltrimethylammonium dichloroiodate, 99%, Thermo Scientific Chemicals
Benzyltrimethylammonium dichloroiodate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyltrimethylammonium dichloroiodate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 268601000
Price (USD)/ Each
430.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
430.00
Add to cart
Benzyltrimethylammonium dichloroiodate, 99%, Thermo Scientific Chemicals
Catalog number268601000
Price (USD)/ Each
430.00
-
Add to cart
Chemical Identifiers
CAS114971-52-7
IUPAC Namebenzyltrimethylazanium; dichloroiodanuide
Molecular FormulaC10H16Cl2IN
InChI KeyPPDJNZTUDFPAHX-UHFFFAOYSA-N
SMILESCl[I-]Cl.C[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
Melting point125°C to 128°C
Appearance (Color)Yellow to light brown
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only