Chemical Identifiers
CAS15956-28-2
IUPAC Nametetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Molecular FormulaC8H12O8Rh2
InChI KeyVOXKIADTKKPBBP-UHFFFAOYSA-N
SMILESC[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
View more