Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, Thermo Scientific Chemicals
Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 268632500
Price (USD)/ Each
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Molecular FormulaC8H12O8Rh2
InChI KeyVOXKIADTKKPBBP-UHFFFAOYSA-N
SMILESC[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
Molecular Weight (g/mol)441.99
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Assay46 % (Rh)
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only