Thermo Scientific Chemicals

Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 268640050
5 kg, Each
Thermo Scientific Chemicals

Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 268640050
5 kg, Each
Quantity
Catalog number: 268640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
546-89-4
IUPAC Name
lithium(1+) acetate
Molecular Formula
C2H3LiO2
InChI Key
XIXADJRWDQXREU-UHFFFAOYSA-M
SMILES
[Li+].CC([O-])=O
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
280°C to 285°C
Water
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text