Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals

Catalog number: 268810025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals

Catalog number: 268810025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 268810025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Appearance (Form)
Liquid
Residue after evaporation
=<0.0002 %
UV
at 210 nm A: =<0.2
UV
at 230 nm A: =<0.02
UV
1 cm cell vs HPLC-grade water

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text