Thermo Scientific Chemicals

5-Methyl-2-furanmethanamine, 98%, Thermo Scientific Chemicals

Catalog number: 268940250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Methyl-2-furanmethanamine, 98%, Thermo Scientific Chemicals

Catalog number: 268940250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 268940250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14003-16-8
IUPAC Name
1-(5-methylfuran-2-yl)methanamine
Molecular Formula
C6H9NO
InChI Key
YSEAGSCGERFGBL-UHFFFAOYSA-N
SMILES
CC1=CC=C(CN)O1
Appearance (Color)
Clear yellow
Titration with HClO4
>=97.5 %
Refractive index
1.4860 to 1.4880 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text