1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, mixture of isomers, Thermo Scientific Chemicals
1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, mixture of isomers, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number 269010010
Price (USD)/ Each
149.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
176.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
149.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
176.00 
Save 26.35 (15%)
Add to cart
1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, mixture of isomers, Thermo Scientific Chemicals
Catalog number269010010
Price (USD)/ Each
149.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
176.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrisodium naphthalene-1,3,6-trisulfonate
Molecular FormulaC10H5Na3O9S3
InChI KeyNJPKYOIXTSGVAN-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=CC(=CC(=C2C=C1)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol)434.29
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<12 %
Appearance (Color)White to yellow to beige
Appearance (Form)Powder
HPLC>=85.0 % (mixture of isomers)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only