4-Pentenoic acid, 99%, Thermo Scientific Chemicals
4-Pentenoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Pentenoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 269020050
Price (USD)/ Each
35.10
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
35.10
Add to cart
4-Pentenoic acid, 99%, Thermo Scientific Chemicals
Catalog number269020050
Price (USD)/ Each
35.10
-
Add to cart
Chemical Identifiers
CAS591-80-0
IUPAC Namepent-4-enoic acid
Molecular FormulaC5H8O2
InChI KeyHVAMZGADVCBITI-UHFFFAOYSA-N
SMILESOC(=O)CCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Specific gravity(25°C) 0.971 to 0.979
Infrared spectrumConforms
Refractive index1.4273 to 1.4303 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only