Ethyl 3-furoate, 99%, Thermo Scientific Chemicals
Ethyl 3-furoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-furoate, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 269030050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS614-98-2
IUPAC Nameethyl furan-3-carboxylate
Molecular FormulaC7H8O3
InChI KeyLOFDXZJSDVCYAS-UHFFFAOYSA-N
SMILESCCOC(=O)C1=COC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4600 to 1.4620 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only