Thermo Scientific Chemicals

Triphenylphosphonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 269091000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Triphenylphosphonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 269091000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 269091000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6399-81-1
IUPAC Name
triphenylphosphane hydrobromide
Molecular Formula
C18H16BrP
InChI Key
CMSYDJVRTHCWFP-UHFFFAOYSA-N
SMILES
Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder and/or chunks
Titration Argentometric
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text