Thermo Scientific Chemicals

Copper phthalocyanine, dye content ca 95%, Thermo Scientific Chemicals

Catalog number: 269150250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Copper phthalocyanine, dye content ca 95%, Thermo Scientific Chemicals

Catalog number: 269150250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 269150250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
147-14-8
IUPAC Name
9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-nonadecaene
Molecular Formula
C32H16CuN8
InChI Key
XCJYREBRNVKWGJ-UHFFFAOYSA-N
SMILES
[Cu]1N2C3=NC4=NC(=NC5=C6C=CC=CC6=C(N=C6N=C(N=C2C2=CC=CC=C32)C2=CC=CC=C62)N15)C1=CC=CC=C41
Appearance (Form)
Crystalline powder
UV spectrum
Complies
Infrared spectrum
Conforms
Appearance (Color)
Dark blue
Titration with Na2S2O3
>=94 % (ex Cu)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text