Chemical Identifiers
CAS147-14-8
IUPAC Name9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-nonadecaene
Molecular FormulaC32H16CuN8
InChI KeyXCJYREBRNVKWGJ-UHFFFAOYSA-N
SMILES[Cu]1N2C3=NC4=NC(=NC5=C6C=CC=CC6=C(N=C6N=C(N=C2C2=CC=CC=C32)C2=CC=CC=C62)N15)C1=CC=CC=C41
View more