Thermo Scientific Chemicals

Potassium tetrafluoroborate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 269220250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium tetrafluoroborate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 269220250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 269220250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14075-53-7
IUPAC Name
potassium tetrafluoroboranuide
Molecular Formula
BF4K
InChI Key
AKEBROIVCDHVSD-UHFFFAOYSA-N
SMILES
[K+].F[B-](F)(F)F
Appearance (Color)
White to light grey
Silicate (as SiO2)
=<0.05 %
Sulfate (SO4)
=<50 ppm
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<30 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text