Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 269551000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS38330-80-2
IUPAC Namepotassium 3-methoxy-3-oxopropanoate
Molecular FormulaC4H5KO4
InChI KeyWWTULTKUWBKVGV-UHFFFAOYSA-M
SMILES[K+].COC(=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only