Thermo Scientific Chemicals

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%, Thermo Scientific Chemicals

Catalog number: 269631000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%, Thermo Scientific Chemicals

Catalog number: 269631000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 269631000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
924-99-2
IUPAC Name
ethyl (2Z)-3-(dimethylamino)prop-2-enoate
Molecular Formula
C7H13NO2
InChI Key
MVUMJYQUKKUOHO-WAYWQWQTSA-N
SMILES
CCOC(=O)C=C/N(C)C
Appearance (Color)
Clear yellow to light orange
Refractive index
1.5105 to 1.5125 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text