Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific Chemicals
Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number 269641000
Price (USD)/ Each
226.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
267.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
226.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
267.00 
Save 40.35 (15%)
Add to cart
Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific Chemicals
Catalog number269641000
Price (USD)/ Each
226.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
267.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethyl 3,3-diethoxypropanoate
Molecular FormulaC9H18O4
InChI KeySIALOQYKFQEKOG-UHFFFAOYSA-N
SMILESCCOC(CC(=O)OCC)OCC
Molecular Weight (g/mol)190.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only