Bis(2-ethylhexyl) sebacate, 97%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) sebacate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(2-ethylhexyl) sebacate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 269920010
Price (USD)/ Each
73.65
Online exclusive
81.30 
Save 7.65 (9%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
81.30 
Save 7.65 (9%)
Add to cart
Bis(2-ethylhexyl) sebacate, 97%, Thermo Scientific Chemicals
Catalog number269920010
Price (USD)/ Each
73.65
Online exclusive
81.30 
Save 7.65 (9%)
-
Add to cart
Chemical Identifiers
CAS122-62-3
IUPAC Name1,10-bis(2-ethylhexyl) decanedioate
Molecular FormulaC26H50O4
InChI KeyVJHINFRRDQUWOJ-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4500 to 1.4550 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only