Thermo Scientific Chemicals

Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals

Catalog number: 269940250
25 g, Each
Thermo Scientific Chemicals

Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals

Catalog number: 269940250
25 g, Each
Quantity
Catalog number: 269940250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
355-43-1
IUPAC Name
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
Molecular Formula
C6F13I
InChI Key
BULLJMKUVKYZDJ-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Appearance (Color)
Clear light yellow to violet
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.3275 to 1.3295 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text