Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals
Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 269941000
Price (USD)/ Each
453.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
453.00
Add to cart
Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals
Catalog number269941000
Price (USD)/ Each
453.00
-
Add to cart
Chemical Identifiers
CAS355-43-1
IUPAC Name1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
Molecular FormulaC6F13I
InChI KeyBULLJMKUVKYZDJ-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to violet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3275 to 1.3295 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only