Thermo Scientific Chemicals

Perfluorobutyl iodide, 99%, Thermo Scientific Chemicals

Catalog number: 269950250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Perfluorobutyl iodide, 99%, Thermo Scientific Chemicals

Catalog number: 269950250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 269950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
423-39-2
IUPAC Name
1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Molecular Formula
C4F9I
InChI Key
PGRFXXCKHGIFSV-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.3275 to 1.3295 (20°C, 589 nm)
Appearance (Color)
Clear purple

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text