Perfluorobutyl iodide, 99%, Thermo Scientific Chemicals
Perfluorobutyl iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluorobutyl iodide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 269951000
Price (USD)/ Each
198.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
198.00
Add to cart
Perfluorobutyl iodide, 99%, Thermo Scientific Chemicals
Catalog number269951000
Price (USD)/ Each
198.00
-
Add to cart
Chemical Identifiers
CAS423-39-2
IUPAC Name1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Molecular FormulaC4F9I
InChI KeyPGRFXXCKHGIFSV-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3275 to 1.3295 (20°C, 589 nm)
Appearance (Color)Clear purple
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only