Thermo Scientific Chemicals

Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals

Catalog number: 269960500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals

Catalog number: 269960500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 269960500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
507-63-1
IUPAC Name
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
Molecular Formula
C8F17I
InChI Key
KWXGJTSJUKTDQU-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Appearance (Form)
Liquid after melting
Appearance (Color)
Clear colorless to yellow to light pink
GC
>=97.5 %
Refractive index
1.3280 to 1.3310 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text