C3 Spacer Phosphoramidite, standard grade, serum vial bottle
Access to customized nucleic acid chemistry for producing oligo therapeutics. Inquire about available services.
Thermo Scientific™
C3 Spacer Phosphoramidite, standard grade, serum vial bottle
C3 Spacer CEP is a phosphoramidite used to incorporate a three carbon spacer arm during oligonucleotide synthesis. Spacers are designedRead more
Have Questions?
Catalog number 27-1795-04
Price (USD)/ 0.25 g
109.00
-
Add to cart
Price (USD)/ 0.25 g
109.00
Add to cart
C3 Spacer Phosphoramidite, standard grade, serum vial bottle
Catalog number27-1795-04
Price (USD)/ 0.25 g
109.00
-
Add to cart
C3 Spacer CEP is a phosphoramidite used to incorporate a three carbon spacer arm during oligonucleotide synthesis. Spacers are designed to add space either within an oligo sequence or between the oligo and a label (e.g. FAM, biotin). The C3 spacer phosphoramidite is used to incorporate a three carbon spacer arm. For the creation of longer spacer arms, multiple sequential additions can be utilized. This product is supplied in a 5 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
• Full chemical name—3-(4,4'-Dimethoxytrityloxy)propyl-1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C33H43N2O5P
• Formula weight—578.7 g/mol
• CAS number—NA!!
• HPLC Purity—≥98.0%
• 31P NMR Purity—≥98.0%; Sum of PIII impurities (141-152ppm) ≤ 0.5%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatOil
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
Product TypeC3 Spacer CEP
Nucleoside TypeNone (Modifier)
Shipping ConditionDry Ice
Purity98%
Quantity0.25 g
Reagent TypePhosphoramidite
GradeStandard
Nucleic Acid TypeNone (Modifier)
Unit Size0.25 g
Contents & Storage
Store in freezer (at or below -20°C).