Informational:Access to customized nucleic acid chemistry for producing oligo therapeutics. Inquire about available services.
Thermo Scientific™

TheraPure™ Ac-rC Phosphoramidite, wide mouth bottle

Catalog number: 27-1805-50
Have Questions?
Thermo Scientific™

TheraPure™ Ac-rC Phosphoramidite, wide mouth bottle

Catalog number: 27-1805-50
Have Questions?
Catalog Number
27-1805-50
Unit Size
50 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
27-1805-5050 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Ac-rC CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 250 mL Opaque bottle with a Wide Mouth neck style, which is compatible with most None (Bulk) of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-acetyl-2'-O-[(tert-butyl)dimethylsilyl]-cytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C47H64N5O9PSi
• Base—rC
• Base Protection—Ac
• Formula weight—902.1 g/mol
• CAS number—121058-88-6
• HPLC Purity—≥99%; M-11 Impurity: ≤0.01%
• 31P NMR Purity—≥99%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%; No single PIII impurity >0.3%

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Powder
For Use With (Equipment)
None (Bulk)
Product Type
Ac-rC CEP
Nucleoside Type
Cytidine (C)
Reagent Features
Fast deprotection
Shipping Condition
Room Temperature
Purity
99%
Reagent Type
Phosphoramidite
Grade
TheraPure™
Nucleic Acid Type
RNA
Product Line
TheraPure™

Contents & Storage

Store in freezer (at or below -20°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text