Informational:Access to customized nucleic acid chemistry for producing oligo therapeutics. Inquire about available services.
Thermo Scientific™

2'-OMe-U Phosphoramidite, standard grade, serum vial bottle

Catalog number: 27-1825-02
Have Questions?
Thermo Scientific™

2'-OMe-U Phosphoramidite, standard grade, serum vial bottle

Catalog number: 27-1825-02
Have Questions?
Catalog Number
27-1825-02
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
27-1825-021 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyl-uridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C40H56N7O9P
• Base—U
• Formula weight—760.8 g/mol
• CAS number—110764-79-9
• HPLC purity—≥98.0%; Sum of non-primary peaks at 140-152ppm: ≤0.5%
31P NMR purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Powder
For Use With (Equipment)
AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type
2'-OMe-U CEP
Nucleoside Type
Uridine (U)
Reagent Features
2'-OMe modified
Shipping Condition
Room Temperature
Purity
98%
Reagent Type
Phosphoramidite
Grade
Standard
Nucleic Acid Type
RNA

Contents & Storage

Store in freezer (at or below -20°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text