Informational:Access to customized nucleic acid chemistry for producing oligo therapeutics. Inquire about available services.
Thermo Scientific™

TheraPure™ iBu-dG Phosphoramidite, serum vial bottle

Catalog number: 27-2034-12
Have Questions?
Thermo Scientific™

TheraPure™ iBu-dG Phosphoramidite, serum vial bottle

Catalog number: 27-2034-12
Have Questions?
Catalog Number
27-2034-12
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
27-2034-121 g
-
27-2034-072 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The TheraPure trademark denotes a highest level of phosphoramidite purity, developed with the highest stringency of of high definiteion analytics and tightly controlled impurity profiles. Ideal for any therapeutic application. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C44H54N7O8P
• Base—dG
• Base Protection—iBu
• Formula weight—839.92 g/mol
• CAS number—93183-15-4
• HPLC Purity—≥99%; M-11 Impurity ≤0.01%; Single critical impurity ≤0.3%; Presence of dA, dC, or T impurity ≤0.03%
• 31P NMR Purity—≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%; No peak(s) above 170 ppm; HMT impurity (∼146ppm): ≤0.10%

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Powder
For Use With (Equipment)
AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type
iBu-dG CEP
Nucleoside Type
Deoxyguanosine (dG)
Shipping Condition
Room Temperature
Purity
99%
Reagent Type
Phosphoramidite
Grade
TheraPure™
Nucleic Acid Type
DNA
Product Line
TheraPure™

Contents & Storage

Store in freezer (at or below -20°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text