(-)-Terpinen-4-ol, 97%, sum of enantiomers
(-)-Terpinen-4-ol, 97%, sum of enantiomers
(-)-Terpinen-4-ol, 97%, sum of enantiomers
(-)-Terpinen-4-ol, 97%, sum of enantiomers
Thermo Scientific Chemicals

(-)-Terpinen-4-ol, 97%, sum of enantiomers

CAS: 20126-76-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 270431000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20126-76-5
IUPAC Name(1R)-4-methyl-1-(propan-2-yl)cyclohex-3-en-1-ol
Molecular FormulaC10H18O
InChI KeyWRYLYDPHFGVWKC-JTQLQIEISA-N
SMILESCC(C)[C@@]1(O)CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-19° to -31° (20°C, 589 nm) (neat)
Infrared spectrumConforms
GC>=96.0 % (sum of enantiomers)
Density(20/4°C) 0.930 to 0.936 g/ml
Refractive index1.4760 to 1.4820 (20°C, 589 nm)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0474265Certificate of AnalysisJul 14, 2025270431000, 270430250
A0461054Certificate of AnalysisMar 13, 2024270431000, 270430250
A0451606Certificate of AnalysisMay 02, 2023270431000, 270430250
A0418029Certificate of AnalysisDec 12, 2022270431000, 270430250
A0397838Certificate of AnalysisDec 12, 2022270431000, 270430250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection