Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
250 mL
5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 270440010
Price (USD)/ Each
142.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
168.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
142.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
168.00 
Save 25.35 (15%)
Add to cart
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals
Catalog number270440010
Price (USD)/ Each
142.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
168.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrimethylamine
Molecular FormulaC3H9N
InChI KeyGETQZCLCWQTVFV-UHFFFAOYSA-N
SMILESCN(C)C
Molecular Weight (g/mol)59.11
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl32 to 34 %
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use