Thermo Scientific Chemicals

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals

Catalog number: 270440250
25 mL, Each
Thermo Scientific Chemicals

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals

Catalog number: 270440250
25 mL, Each
Quantity
Catalog number: 270440250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-50-3, 64-17-5
Titration with HCl
32 to 34 %
Appearance (Color)
Clear colorless
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text