1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number 270490025
Price (USD)/ Each
177.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
209.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
177.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
209.00 
Save 31.35 (15%)
Add to cart
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
Catalog number270490025
Price (USD)/ Each
177.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
209.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.1 %
Chromium (Cr)=<0.1 ppm
Copper (Cu)=<0.1 ppm
Stabilizer5 to 30 ppm BHT
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only