Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 270510050
5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 270510050
5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 270510050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7791-20-0
IUPAC Name
nickel(2+) hexahydrate dichloride
Molecular Formula
Cl2H12NiO6
InChI Key
LAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
Appearance (Color)
Green
Appearance (Form)
Crystalline powder or crystals
Cobalt (Co)
=<60 ppm
Iron (Fe)
=<20 ppm
Zinc (Zn)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text