Nickel(II) chloride hexahydrate, 98+%
Nickel(II) chloride hexahydrate, 98+%
Nickel(II) chloride hexahydrate, 98+%
Nickel(II) chloride hexahydrate, 98+%
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 98+%

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 270512500
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Appearance (Form)Crystalline powder or crystals
Cobalt (Co)=<60 ppm
Iron (Fe)=<20 ppm
Zinc (Zn)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link