Copper(I) chloride, 97+%, extra pure, Thermo Scientific Chemicals
Copper(I) chloride, 97+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(I) chloride, 97+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
1 kg
Packaging:
Glass bottle
Catalog number 270520025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameλ¹-copper(1+) chloride
Molecular FormulaClCu
InChI KeyOXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES[Cl-].[Cu+]
Molecular Weight (g/mol)99.00
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric97.0 to 103.0 %
Insoluble matter=<0.05 % (in acid)
Appearance (Color)Light green to grey
Appearance (Form)Crystalline powder
Iron (Fe)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only