Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970
Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970
Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970
Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970
Thermo Scientific Chemicals

Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970

CAS: 10101-98-1 | H14NiO11S | 280.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 270552500
Price (USD)
100.00
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
100.00
Each
Add to cart
Chemical Identifiers
CAS10101-98-1
IUPAC Namenickel(2+) heptahydrate sulfate
Molecular FormulaH14NiO11S
InChI KeyOGKAGKFVPCOHQW-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<10 ppm
Appearance (Color)Green to dark green
Titration Complexometric20 to 21 % (Ni)
Arsenic (As)=<10 ppm
Cadmium (Cd)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link