Thermo Scientific Chemicals

Alginic acid

Catalog number: 270581000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Alginic acid

Catalog number: 270581000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 270581000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-32-7
Molecular Formula
(C6H8O7)A(C6H8O7)B
SMILES
O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O
Molecular Weight (g/mol)
NaN
Appearance (Color)
White to yellow to light beige
Infrared spectrum
Conforms
Loss on drying
=<15 % (2 g, 105°C, 4 hrs)
Titration excess NaOH
19 to 25 % (Carboxyl groups) On dried substance
Sulfated ash
=<8 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text