Triethylamine hydrochloride, 99+%, Thermo Scientific Chemicals
Triethylamine hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylamine hydrochloride, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 270670010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namehydrogen triethylamine chloride
Molecular FormulaC6H16ClN
InChI KeyILWRPSCZWQJDMK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCN(CC)CC
Molecular Weight (g/mol)137.65
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 % (On anhydrous substance)
pH4.5 to 6.5 (1 % w/v solution)
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only