Thermo Scientific Chemicals

Triethylamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 270672500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethylamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 270672500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 270672500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-68-7
IUPAC Name
hydrogen triethylamine chloride
Molecular Formula
C6H16ClN
InChI Key
ILWRPSCZWQJDMK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCN(CC)CC
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
99.0 to 101.0 % (On anhydrous substance)
pH
4.5 to 6.5 (1 % w/v solution)
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text