1x1 image pixel for data collection
Fehling A solution
Fehling A solution
Fehling A solution
Thermo Scientific Chemicals

Fehling A solution

CAS: 7758-98-7 | CuO4S | 159.602 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Catalog number 270800010
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
55.10
Save 10.45 (19%)
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
55.10
Save 10.45 (19%)
Each
Add to cart
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Solution contains69.2 g CuSO4.5H2O/l H2O
Appearance (Color)Clear blue
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link