Thermo Scientific Chemicals

Methyl-beta-D-glucopyranoside hemihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 270920250
Each
Thermo Scientific Chemicals

Methyl-beta-D-glucopyranoside hemihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 270920250
Each
Catalog number: 270920250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7000-27-3
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Formula
C7H14O6
InChI Key
HOVAGTYPODGVJG-XUUWZHRGSA-N
SMILES
CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Appearance (Form)
Crystalline powder
Water
=<5.4 % (K.F.)
Appearance (Color)
White
HPLC
>=98.5 % (ELSD)
Solubility
(10 % in water) Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text