Thermo Scientific Chemicals

2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%, Thermo Scientific Chemicals

Catalog number: 270930100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%, Thermo Scientific Chemicals

Catalog number: 270930100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 270930100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
572-09-8
IUPAC Name
[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Molecular Formula
C14H19BrO9
InChI Key
CYAYKKUWALRRPA-HAGGZJDPNA-N
SMILES
CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
Titration Argentometric
>=98.0 %
Specific optical rotation
+188° to +202° (20°C, 589 nm) (c=2, CHCl3)
Appearance (Form)
Powder
Appearance (Color)
White to beige to light grey
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text