Chemical Identifiers
CAS2595-05-3
IUPAC Name(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular FormulaC12H20O6
InChI KeyKEJGAYKWRDILTF-VVULQXIFSA-N
SMILESCC1(C)OC[C@@H](O1)[C@H]1O[C@@H]2OC(C)(C)O[C@@H]2[C@@H]1O
View more