Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 271060100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 271060100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 271060100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10039-32-4
IUPAC Name
disodium dodecahydrate hydrogen phosphate
Molecular Formula
H25Na2O16P
InChI Key
DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
Appearance (Form)
Crystals
Arsenic (As)
=<0.5 ppm
Cobalt (Co)
=<5 ppm
Copper (Cu)
=<5 ppm
Magnesium (Mg)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text