Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals
Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 271070025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS6132-02-1
IUPAC Namedisodium decahydrate carbonate
Molecular FormulaCH20Na2O13
InChI KeyXYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration with HCl99.0 to 102.0 %
Iron (Fe)=<4 ppm
Chloride (Cl)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only